Learn More
Medchemexpress LLC Hydroquinidine | 1435-55-8 | 99.97% | 5 G
SDP
Catalog No. 5000407455
Encompass_Preferred
Hydroquinidine (Dihydroquinidine) is a potent ion channel blocker that exhibits strong anti-cancer activity on colon, pancreatic, and hepatocellular cancer cells. It also prolongs the QT interval and demonstrates antiarrhythmic efficacy.
  • Potent ion channel blocker
  • Strong anti-cancer activity on colon, pancreatic, and hepatocellular cancer cells
  • Prolongs the QT interval
  • Antiarrhythmic efficacy
  • For research use only
Catalog No. 50-004-07455 Supplier Medchemexpress LLC Supplier No. HYB09975G
Hydroquinidine (Dihydroquinidine) is a potent ion channel blocker that exhibits strong anti-cancer activity on colon, pancreatic, and hepatocellular cancer cells. It also prolongs the QT interval and demonstrates antiarrhythmic efficacy.
  • Potent ion channel blocker
  • Strong anti-cancer activity on colon, pancreatic, and hepatocellular cancer cells
  • Prolongs the QT interval
  • Antiarrhythmic efficacy
  • For research use only