Learn More
Apexbio Technology LLC HYDROQUINIDINE-200MG
SDP
Catalog No. 5000570041
Encompass_Preferred
Hydroquinidine (CAS No 1435-55-8) is utilized in biomedical research primarily for its role as a cardioactive compound originating from the bark of the Cinchona tree It operates by modulating cardiac ion channels particularly targeting sodium and potassium channels to stabilize myocardial electrical activity In vitro its effective concentrations are typically observed in the nanomolar to low micromolar range This compound is frequently utilized in studies involving cardiac cell models and has applications in animal studies for examining cardiac arrhythmias It is also employed in drug screening processes aimed at discovering and evaluating new antiarrhythmic agents Experimental concentrations typically depend on the specific research design ensuring adaptability across varied experimental setups
Catalog No. 50-005-70041 Supplier Apexbio Technology LLC Supplier No. C6712200
Hydroquinidine (CAS No 1435-55-8) is utilized in biomedical research primarily for its role as a cardioactive compound originating from the bark of the Cinchona tree It operates by modulating cardiac ion channels particularly targeting sodium and potassium channels to stabilize myocardial electrical activity In vitro its effective concentrations are typically observed in the nanomolar to low micromolar range This compound is frequently utilized in studies involving cardiac cell models and has applications in animal studies for examining cardiac arrhythmias It is also employed in drug screening processes aimed at discovering and evaluating new antiarrhythmic agents Experimental concentrations typically depend on the specific research design ensuring adaptability across varied experimental setups