Learn More
Medchemexpress LLC Hydroquinidine (Dihydroquinidine) | 1435-55-8 | 99.9% | 50 G
SDP
Catalog No. 5000407553
Encompass_Preferred
Hydroquinidine (Dihydroquinidine) is a potent ion channel blocker that demonstrates strong anti-cancer activity against colon, pancreatic, and hepatocellular cancer cells. It also prolongs the QT interval and possesses antiarrhythmic efficacy.
  • Exhibits anti-clonogenic and anti-tumorigenic activities in pancreatic cancer cells in vitro.
  • Gene expression profiling shows changes in CDKN1A, CDKN1B, PUMA, BIRC5, AKT, MKI67, PCNA, and CCNE1 in various cancer cell lines.
  • Investigated for Brugada Syndrome in a Phase 3 clinical trial.
Catalog No. 50-004-07553 Supplier Medchemexpress LLC Supplier No. HYB099750G
Hydroquinidine (Dihydroquinidine) is a potent ion channel blocker that demonstrates strong anti-cancer activity against colon, pancreatic, and hepatocellular cancer cells. It also prolongs the QT interval and possesses antiarrhythmic efficacy.
  • Exhibits anti-clonogenic and anti-tumorigenic activities in pancreatic cancer cells in vitro.
  • Gene expression profiling shows changes in CDKN1A, CDKN1B, PUMA, BIRC5, AKT, MKI67, PCNA, and CCNE1 in various cancer cell lines.
  • Investigated for Brugada Syndrome in a Phase 3 clinical trial.