Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroquinidine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. H075225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H075225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H075225G Supplier TCI America Supplier No. H075225G
Add to Cart
Add to Cart

Specifications

CAS 1476-98-8
Color White
Molecular Formula C20H27ClN2O2
Quantity 25 g
MDL Number MFCD00135598
Synonym Dihydroquinidine Hydrochloride
InChI Key MULXTQKDWYBJMO-VJAUXQICSA-N
SMILES CCC1CN2CCC1CC2C(C3=C4C=C(C=CC4=NC=C3)OC)O.Cl
IUPAC Name (S)-[(2R,4S,5R)-5-ethyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;hydrochloride
Molecular Weight (g/mol) 362.898
PubChem CID 16211709
Formula Weight 362.90
Percent Purity ≥98.0% (T,HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Hydroquinidine Hydrochloride
Show More Show Less
RTECSNumber MX3018000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.