Learn More
Selleck Chemical LLC Hydroquinone-100mg
SDP
Catalog No. 502389684
Encompass_Preferred
Hydroquinone (Quinol, 1,4-Benzenediol, 1,4-Dihydroxybenzene, HQ) is a Melanin Synthesis Inhibitor. The physiologic effect of hydroquinone is by means of Depigmenting Activity. It also posesses antioxidant properties that can cause toxicity in several organs, notably the kidney.
Catalog No. 50-238-9684 Supplier Selleck Chemical LLC Supplier No. S4580100MG
Hydroquinone (Quinol, 1,4-Benzenediol, 1,4-Dihydroxybenzene, HQ) is a Melanin Synthesis Inhibitor. The physiologic effect of hydroquinone is by means of Depigmenting Activity. It also posesses antioxidant properties that can cause toxicity in several organs, notably the kidney.