Learn More
Selleck Chemical LLC Hydroquinone S4580-500mg
SDP
Catalog No. 503111495
Encompass_Preferred
Hydroquinone (Quinol 1 4-Benzenediol 1 4-Dihydroxybenzene HQ) is a Melanin Synthesis Inhibitor The physiologic effect of hydroquinone is by means of Depigmenting Activity It also posesses antioxidant properties that can cause toxicity in several organs notably the kidney This product is a hazardous chemical (acute toxicity/flammable/skin corrosive) Please use it while wearing a protective face mask gloves and clothing
Catalog No. 50-311-1495 Supplier Selleck Chemical LLC Supplier No. S4580500MG
Hydroquinone (Quinol 1 4-Benzenediol 1 4-Dihydroxybenzene HQ) is a Melanin Synthesis Inhibitor The physiologic effect of hydroquinone is by means of Depigmenting Activity It also posesses antioxidant properties that can cause toxicity in several organs notably the kidney This product is a hazardous chemical (acute toxicity/flammable/skin corrosive) Please use it while wearing a protective face mask gloves and clothing