Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroquinone (Crystalline/Laboratory), Fisher Chemical

Catalog No. H329500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
H329-500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H329-500

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 123-31-9
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
MDL Number MFCD00002339
InChI Key QIGBRXMKCJKVMJ-UHFFFAOYSA-N
Synonym hydroquinone, 1,4-benzenediol, quinol, 1,4-dihydroxybenzene, p-benzenediol, 4-hydroxyphenol, p-hydroquinone, p-hydroxyphenol, p-dihydroxybenzene, benzoquinol
PubChem CID 785
ChEBI CHEBI:17594
IUPAC Name benzene-1,4-diol
SMILES C1=CC(=CC=C1O)O

Specifications

Melting Point 170°C
Color White
Density 1.320g/cm³
pH 3.75
Boiling Point 285°C
Packaging Poly Bottle
Linear Formula 1, 4-HOC6H4OH
Quantity 500 g
UN Number 2662
Vapor Density 3.8g/mol
Formula Weight 110.11
Grade Laboratory
Physical Form Solid
Chemical Name or Material Hydroquinone
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation Class 6.1: Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.