Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydroquinone diacetate (1,4-diacetoxybenzene) | 1205-91-0 | MFCD00011643 | 98.3% | 194.19 g/mol | C10H10O4 | 100 MG
SDP

Catalog No. 5000227883
Encompass_Preferred

Hydroquinone diacetate is the diacetyl ester of hydroquinone supplied as a solid powder for research use. It is used as a synthetic intermediate in organic and medicinal chemistry and is also applied in formulation studies. The manufacturer provides a datasheet and SDS with analytical data, storage, and handling recommendations.

  • Solid powder form suitable for laboratory use.
  • High purity: 98.3% by manufacturer analysis.
  • Molecular weight 194.19 g/mol and formula C10H10O4.
  • Recommended storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C or -20°C.
  • Intended for research and synthesis applications as a building block.

Catalog No. 50-002-27883 Supplier Medchemexpress LLC Supplier No. HY118292100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydroquinone diacetate is the diacetyl ester of hydroquinone supplied as a solid powder for research use. It is used as a synthetic intermediate in organic and medicinal chemistry and is also applied in formulation studies. The manufacturer provides a datasheet and SDS with analytical data, storage, and handling recommendations.

  • Solid powder form suitable for laboratory use.
  • High purity: 98.3% by manufacturer analysis.
  • Molecular weight 194.19 g/mol and formula C10H10O4.
  • Recommended storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C or -20°C.
  • Intended for research and synthesis applications as a building block.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.