Learn More
Medchemexpress LLC Hydroquinone diacetate | 1205-91-0 | MFCD00011643 | ≥96.0% | 194.19 g/mol | C10H10O4 | 5 MG
SDP
Catalog No. 5000227461
Encompass_Preferred
Hydroquinone diacetate is the diacetate ester of hydroquinone used as an intermediate in organic and medicinal chemistry and is listed by the manufacturer as commonly used in skin-lightening formulation research. It is supplied as a white to off-white solid (formula C10H10O4, MW 194.19 g/mol) in small laboratory pack sizes and requires refrigerated or frozen storage depending on preparation.
  • Diacetate-protected hydroquinone suitable for synthetic transformations.
  • Commonly used as an intermediate in skin-lightening formulation research.
  • White to off-white solid with defined molecular formula and weight.
  • Supplied in small research pack sizes for laboratory use.
  • Store under recommended refrigerated or frozen conditions to preserve stability.
Catalog No. 50-002-27461 Supplier Medchemexpress LLC Supplier No. HY1182925MG
Hydroquinone diacetate is the diacetate ester of hydroquinone used as an intermediate in organic and medicinal chemistry and is listed by the manufacturer as commonly used in skin-lightening formulation research. It is supplied as a white to off-white solid (formula C10H10O4, MW 194.19 g/mol) in small laboratory pack sizes and requires refrigerated or frozen storage depending on preparation.
  • Diacetate-protected hydroquinone suitable for synthetic transformations.
  • Commonly used as an intermediate in skin-lightening formulation research.
  • White to off-white solid with defined molecular formula and weight.
  • Supplied in small research pack sizes for laboratory use.
  • Store under recommended refrigerated or frozen conditions to preserve stability.