Learn More
Medchemexpress LLC Hydroquinone diacetate | 1205-91-0 | MFCD00011643 | 98.3% | 194.19 g/mol | C10H10O4 | 1 G
SDP
Catalog No. 5000227462
Encompass_Preferred
Hydroquinone diacetate is the diacetyl ester of hydroquinone used as a chemical intermediate in organic and medicinal chemistry and in topical formulations such as skin lighteners. It is supplied as a powder and in solution formats for research and development.
  • Used as a chemical intermediate in synthesis and medicinal chemistry.
  • Available as a powder and as pre-made solutions for convenience.
  • High purity suitable for research and development applications.
  • Stable when stored cold; follow documented storage recommendations.
  • SDS, data sheet, and certificate of analysis available for handling and quality verification.
Catalog No. 50-002-27462 Supplier Medchemexpress LLC Supplier No. HY1182921G
Hydroquinone diacetate is the diacetyl ester of hydroquinone used as a chemical intermediate in organic and medicinal chemistry and in topical formulations such as skin lighteners. It is supplied as a powder and in solution formats for research and development.
  • Used as a chemical intermediate in synthesis and medicinal chemistry.
  • Available as a powder and as pre-made solutions for convenience.
  • High purity suitable for research and development applications.
  • Stable when stored cold; follow documented storage recommendations.
  • SDS, data sheet, and certificate of analysis available for handling and quality verification.