Learn More
Medchemexpress LLC Hydroquinone diacetate | 1205-91-0 | MFCD00011643 | 98.3% | 194.19 g/mol | C10H10O4 | 1 ML
SDP
Catalog No. 5000227689
Encompass_Preferred
Hydroquinone diacetate (1,4-diacetoxybenzene) is the diacetylated derivative of hydroquinone used as an intermediate in organic and medicinal chemistry and as an ingredient in topical formulations. The product is supplied as a ready-to-use solution for research, screening, or synthetic use.
  • Ready-to-use 10 mM solution in DMSO for biochemical assays and screening.
  • High purity (≈98.3%) suitable for research applications.
  • Molecular weight 194.19 g/mol; chemical formula C10H10O4.
  • Solid form stable under frozen storage; limited stability in solvent-store cold.
  • Useful as a synthetic intermediate and reagent in medicinal chemistry.
Catalog No. 50-002-27689 Supplier Medchemexpress LLC Supplier No. HY11829210MM 1ML
Hydroquinone diacetate (1,4-diacetoxybenzene) is the diacetylated derivative of hydroquinone used as an intermediate in organic and medicinal chemistry and as an ingredient in topical formulations. The product is supplied as a ready-to-use solution for research, screening, or synthetic use.
  • Ready-to-use 10 mM solution in DMSO for biochemical assays and screening.
  • High purity (≈98.3%) suitable for research applications.
  • Molecular weight 194.19 g/mol; chemical formula C10H10O4.
  • Solid form stable under frozen storage; limited stability in solvent-store cold.
  • Useful as a synthetic intermediate and reagent in medicinal chemistry.