Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydroquinone diacetate | 1205-91-0 | MFCD00011643 | 98.3% | 194.19 g/mol | C10H10O4 | 10 MG
SDP

Catalog No. 5000228523
Encompass_Preferred

Hydroquinone diacetate is an acetylated derivative of hydroquinone used as an intermediate in organic and medicinal chemistry. It is supplied as a solid powder for research applications and may be provided as a solution in DMSO for screening or formulation work.

  • White to off-white solid powder.
  • Molecular weight 194.19 g/mol.
  • Purity about 98.3% by GC.
  • Recommended storage: powder at -20°C or 4°C depending on shelf life.
  • Common uses include organic synthesis intermediates and skin-lightening research.

Catalog No. 50-002-28523 Supplier Medchemexpress LLC Supplier No. HY11829210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydroquinone diacetate is an acetylated derivative of hydroquinone used as an intermediate in organic and medicinal chemistry. It is supplied as a solid powder for research applications and may be provided as a solution in DMSO for screening or formulation work.

  • White to off-white solid powder.
  • Molecular weight 194.19 g/mol.
  • Purity about 98.3% by GC.
  • Recommended storage: powder at -20°C or 4°C depending on shelf life.
  • Common uses include organic synthesis intermediates and skin-lightening research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.