Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydroquinone diacetate | 1205-91-0 | MFCD00011643 | 98.3% | 194.19 g/mol | C10H10O4 | 50 MG
SDP

Catalog No. 5000227884
Encompass_Preferred

Hydroquinone diacetate is an acetylated derivative of hydroquinone used in organic and medicinal chemistry. Supplied as a high-purity research chemical, it is used in synthetic applications and as an ingredient in some skin-lightening formulations.

  • Acetylated hydroquinone derivative used in organic and medicinal chemistry.
  • High purity (98.3%) suitable for research applications.
  • Molecular formula C10H10O4; molecular weight ≈ 194.19 g/mol.
  • White to off-white solid; typically supplied as a powder.
  • Available in small research quantities (for example, 50 mg) and as pre-made DMSO solution for reconstitution.
  • CAS number 1205-91-0 for unambiguous identification.

Catalog No. 50-002-27884 Supplier Medchemexpress LLC Supplier No. HY11829250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydroquinone diacetate is an acetylated derivative of hydroquinone used in organic and medicinal chemistry. Supplied as a high-purity research chemical, it is used in synthetic applications and as an ingredient in some skin-lightening formulations.

  • Acetylated hydroquinone derivative used in organic and medicinal chemistry.
  • High purity (98.3%) suitable for research applications.
  • Molecular formula C10H10O4; molecular weight ≈ 194.19 g/mol.
  • White to off-white solid; typically supplied as a powder.
  • Available in small research quantities (for example, 50 mg) and as pre-made DMSO solution for reconstitution.
  • CAS number 1205-91-0 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.