Learn More
Specifications
Specifications
| Color | Peach |
| CAS Max % | 99, 1 |
| Identification | Passes Test |
| Packaging | Amber Glass |
| Decomposition Information | Carbon monoxide; Carbon dioxide |
| Linear Formula | 1,4-HOC6H4OH |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Solubility Information | Soluble in water |
| Formula Weight | 110.11 |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement May cause an allergic skin reaction. Causes serious eye irritation. Suspected of causing genetic defects. Suspected of causing cancer. Harmful to aquatic life. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Avoid breathing mist. Wear protective gloves, eye protection. Wash exposed skin thoroughly after handling. Contaminated work clothing must not be allowed out of the workplace. Avoid release to the environment. If exposed or concerned: Get medical advice/attention. If on skin: Wash with plenty of soap and water. If skin irritation or rash occurs: Get medical advice/attention. Take off contaminated clothing and wash it before reuse. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. Store locked up. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.