Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroquinone-O,O'-diacetic acid, 98%

Catalog No. AAB2264506
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 2245-53-6 | C10H10O6 | 226.18 g/mol

Hydroquinone-O,O'-diacetic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydroquinone-O,O′-diacetic acid is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2245-53-6
Molecular Formula C10H10O6
Molecular Weight (g/mol) 226.18
MDL Number MFCD00016816
InChI Key DNXOCFKTVLHUMU-UHFFFAOYSA-N
Synonym 2,2'-1,4-phenylenebis oxy diacetic acid, 1,4-phenylenedioxydiacetic acid, hydroquinone-o,o'-diacetic acid, 1,4-dicarboxymethoxybenzene, hydroquinone-o,o-diacetic acid, acetic acid, p-phenylenedioxy di, acetic acid, 2,2'-1,4-phenylenebis oxy bis, 2-4-carboxymethoxy phenoxy acetic acid
PubChem CID 75251
IUPAC Name 2-[4-(carboxymethoxy)phenoxy]acetic acid
SMILES OC(=O)COC1=CC=C(OCC(O)=O)C=C1

Specifications

Melting Point ∼255°C (decomposition)
Quantity 5 g
Beilstein 1984454
Solubility Information Slightly soluble in water.
Formula Weight 226.19
Percent Purity 98%
Chemical Name or Material Hydroquinone-O,O'-diacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-834-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.