Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroquinone, USP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18603272 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-272 2.5 kg Amber Glass Bottle
18-603-270 125 g Amber Glass Bottle
18-603-271 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-603-272 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HY1132.5KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HY113, 123-31-9, C6H6O2

Spectrum™ Chemical Hydroquinone, USP is an effective topical medicine used to treat dark colored patches, freckles, and spots on the skin that appear due to sun exposure. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 123-31-9
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.11
InChI Key QIGBRXMKCJKVMJ-UHFFFAOYSA-N
IUPAC Name benzene-1,4-diol
SMILES OC1=CC=C(O)C=C1

Specifications

CAS Max % 100%
Ignition Residue 0.5%
Melting Range 172°-174°C
Packaging Amber Glass Bottle
Quantity 2.5 kg
UN Number UN3077
Percent Purity 99 to 100.5%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.