Learn More
Dexsil Corporation HYDROSCOUT REAGENT 40 TEST/BOX
Catalog No. NC9479805
Encompass
Hydroscout, Quantification of Water in Several Matrices, Analyte: Water, Matrix: Waste Oil, Industrial Oils, Lubricating Oils, Organic Liquids, Soil, Detection Method: Quantitative Calcium Hydride Reaction, High Range: 1500 ppm - 100% , Low
Catalog No. NC9479805 Supplier Dexsil Corporation Supplier No. HSORP40
Hydroscout, Quantification of Water in Several Matrices, Analyte: Water, Matrix: Waste Oil, Industrial Oils, Lubricating Oils, Organic Liquids, Soil, Detection Method: Quantitative Calcium Hydride Reaction, High Range: 1500 ppm - 100% , Low