Learn More
MSC HYDROSHCK WTRPR ST WRK BT
Catalog No. 5000781207
Encompass
The Lehigh Outfitters work boot is designed for demanding environments, offering superior protection and comfort. It features electric shock-resistance, waterproofing, and dielectric properties, making it suitable for a wide range of industrial applications.
  • Footwear type: Work boot
  • Gender: Unisex
  • Men's size: 3
  • Upper material: Rubber
  • Toe type: Steel
  • Primary color: Green
  • Resistance features: Electric shock-resistant, waterproof, dielectric
  • Meets ASTM F2413 standards
  • Height: 12 in
Catalog No. 50-007-81207 Supplier MSC Supplier No. 50535491
The Lehigh Outfitters work boot is designed for demanding environments, offering superior protection and comfort. It features electric shock-resistance, waterproofing, and dielectric properties, making it suitable for a wide range of industrial applications.
  • Footwear type: Work boot
  • Gender: Unisex
  • Men's size: 3
  • Upper material: Rubber
  • Toe type: Steel
  • Primary color: Green
  • Resistance features: Electric shock-resistant, waterproof, dielectric
  • Meets ASTM F2413 standards
  • Height: 12 in