Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DigiLab HydroShear Plus Hydrodynamic DNA Shearing Instrument

Catalog No. HSP000001
Change view
Click to view available options
For Use With (Equipment):
DNA Sequencers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
HSP000001 DNA Sequencers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HSP000001 Supplier DigiLab Supplier No. HSP000001

missing translation for 'validMainframeMsg'

Shears DNA to produce long fragments. Store specific protocols for shearing different size DNA strands.

  • Automated, multi-port valve in order to allow hands-free multiple washing
  • Fragments DNA sample volumes, as small as 40μL
  • Achieves consistent results across multiple users
  • Generates a random, representative collection of fragments
  • 110V
  • Color: White
  • Shape: Rectangular

Warranty and Services

1 year

Specifications

For Use With (Application) Genomic DNA Sequencing
For Use With (Equipment) DNA Sequencers
Includes One Waste Bottle, Power Cord, Laptop, Software, Shearing Assemblies, Tool Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.