Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HydroSpec™ Coulometric CG, Ricca Chemical
SDP

Catalog No. RK130000110 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
110 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RK130000110 110 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RK130000110 Supplier Ricca Chemical Company Supplier No. RK130000110C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.

Specifications

Chemical Name or Material Coulometric CG
CAS Min % 5, 10
CAS Max % 20, 10, 20
CAS 7446-09-5, 288-32-4, 67-56-1, 111-90-0, 68007-08-9
Physical Form Liquid
Quantity 110 mL
Packaging Glass Bottle
UN Number UN1230
Specific Gravity 0.93
Grade Karl Fischer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.