Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HydroSpec™ Titrant K5, Ricca Chemical
SDP

Catalog No. p-7160822 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K2700001C 1 L
K2700006X1C 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. K2700001C Supplier Ricca Chemical Company Supplier No. RK2700001C
This item is not returnable. View return policy

Achieve high performance and reliable results. Hydrospec™ supports a wide range of sample types that help you produce the fast, reliable, and reproducible results you are looking for.

  • Reliable, Safe, Pyridine-Free
  • Available in 500 mL and 1 L sizes
  • 1 L Bottles have the Industry Standard GL45 Neck Finish for Convenient Connection
  • Stable Formulations and Comprehensive Chemical Compatibility
  • 5 Year Shelf Life for Most Reagents
  • Technical Support Every Step of the Way
  • Tight Specifications for More Accurate Results and Low Lot-to-Lot Variability
  • Serving Scientists Successfully for Over 45 Years

Specifications

CAS Min % 90, 10
CAS Max % 90, 10
CAS 7553-56-2, 107-98-2
Quantity 1 L
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.