Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrous Benzoyl Peroxide, USP, 70-80%, Spectrum™ Chemical
SDP

Catalog No. 18603281 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
1 kg
10 kg
12 kg
45 kg
Packaging:
Poly Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-281 1 kg Poly Bottle
18-603-282 25 g Poly Bottle
18-603-276 125 g Poly Bottle
18-603-277 500 g Poly Bottle
18-603-280 10 kg Poly Bottle
18-603-278 12 kg Poly Bottle
18-603-279 45 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-603-281 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. BE1561KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

BE156, 94-36-0, C14H10O4 (Anhydrous)

Spectrum™ Chemical Hydrous Benzoyl Peroxide, 75 Percent, USP is used to treat mild to moderate acne. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 94-36-0 , 7732-18-5
Molecular Formula C14H10O4
Molecular Weight (g/mol) 242.23
InChI Key OMPJBNCRMGITSC-UHFFFAOYSA-N
IUPAC Name benzoyl benzenecarboperoxoate
SMILES O=C(OOC(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Max % 75%, 25%
Packaging Poly Bottle
Quantity 1 kg
UN Number UN3104
Percent Purity 70 to 80%
Grade USP
Physical Form Slurry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.