Learn More
AIR CONTROL INC Hydrovoid Desiccator Storage
SDP
Catalog No. NC3311177
Encompass_Preferred
Air Control, Inc. Clean Storage Dessicator Cabinets (or dry boxes) areare designed for the protection and storage of moisture and particulate sensitive semiconductor parts; microelectronic devices; biological mixtures; and sensitive processes. By introducing and maintaining an inert gas (nitrogen) to a near-tight environment, stored parts are not subjected to damage caused by oxidation or particulate contamination. Many sizes and custom configurations are available. Other materials include amber acrylic, clear PVC, anti-static acylic, and solid white polypropylene.
Catalog No. NC3311177 Supplier AIR CONTROL INC Supplier No. 43312
Add to Cart
Add to Cart
Air Control, Inc. Clean Storage Dessicator Cabinets (or dry boxes) areare designed for the protection and storage of moisture and particulate sensitive semiconductor parts; microelectronic devices; biological mixtures; and sensitive processes. By introducing and maintaining an inert gas (nitrogen) to a near-tight environment, stored parts are not subjected to damage caused by oxidation or particulate contamination. Many sizes and custom configurations are available. Other materials include amber acrylic, clear PVC, anti-static acylic, and solid white polypropylene.