Learn More
Abcam 8-Hydro x ypyrene-1,3,6-trisulfonic acid trisodium salt, 50MG
Catalog No. 50996229
Encompass_Preferred
MW 524.4 Da. Fluorescent pH indicator with fluorescent emission change in the physiological pH range. Used as a dye in optical sensor applications. Highly water-soluble and membrane-impermeant with a pKa of 7.3 in aqueous buffers.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-229 Supplier Abcam Supplier No. AB27530750MG
MW 524.4 Da. Fluorescent pH indicator with fluorescent emission change in the physiological pH range. Used as a dye in optical sensor applications. Highly water-soluble and membrane-impermeant with a pKa of 7.3 in aqueous buffers.
The product is subject to the following: Abcam Restricted Use Statement