Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxocobalamin, Spectrum™ Chemical
SDP

Catalog No. 18620815 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-815 25 g
18-620-813 1 g
18-620-814 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-620-815 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H139725GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synonym: Coalpha-[Alpha-(5,6-Dimethylbenzimidazolyl)]-CoBeta-Hydroxocobamide, Hydroxocobalamin Acetate

Hydroxocobalamin, also known as hydroxycobalamin, is a natural form of vitamin B12 and member of the cobalamin family of compounds. It is used to treat vitamin deficiency and for treatment for cyanide poisoning. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.se.

Chemical Identifiers

CAS 13422-51-0

Specifications

Quantity 25 g
Molecular Weight (g/mol) 1346.36
Percent Purity 95 to 102%
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Hydroxocobalamin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.