Learn More
Sigma Aldrich Fine Chemicals Biosciences Hydroxocobalamn acetate v250MG
Catalog No. 502897246
Encompass
Hydroxocobalamin is an analog of vitamin B12 with OH as an alkyl group. Vitamin B12 cobalamin refers to a group of chemically-related cobalt containing molecules.
Catalog No. 50-289-7246 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H8017250MG
Hydroxocobalamin is an analog of vitamin B12 with OH as an alkyl group. Vitamin B12 cobalamin refers to a group of chemically-related cobalt containing molecules.