Learn More
Cayman Chemical Hydroxy Bosentan, Purity: ≥98%, CAS: 253688-60-7, 5 mg
SDP
Catalog No. 502987770
Encompass_Preferred
A metabolite of bosentan; produced by CYP3A4 and CYP2C9
Catalog No. 50-298-7770 Supplier Cayman Chemical Supplier No. 223665 MG
A metabolite of bosentan; produced by CYP3A4 and CYP2C9