Learn More
Medchemexpress LLC Hydroxy celecoxib | 170571-00-3 | 98.7% | C17H14F3N3O3S | 10MG
SDP
Catalog No. 5000262057
Encompass_Preferred
Hydroxy celecoxib is a celecoxib derivative that functions as a PI3K/Akt signaling activator and has been reported to promote epithelial repair. It is supplied as a high-purity solid research compound intended for in vitro cellular and pharmacological studies; soluble in DMSO and packaged in milligram quantities.
  • Activates PI3K/Akt signaling and promotes epithelial repair.
  • High purity (~98.7%) appropriate for research use.
  • White to off-white solid with defined storage recommendations.
  • Soluble in DMSO (100 mg/mL); ultrasonic assistance may be needed.
  • Available in small mg package sizes suitable for laboratory experiments.
Catalog No. 50-002-62057 Supplier Medchemexpress LLC Supplier No. HY14552610MG
Hydroxy celecoxib is a celecoxib derivative that functions as a PI3K/Akt signaling activator and has been reported to promote epithelial repair. It is supplied as a high-purity solid research compound intended for in vitro cellular and pharmacological studies; soluble in DMSO and packaged in milligram quantities.
  • Activates PI3K/Akt signaling and promotes epithelial repair.
  • High purity (~98.7%) appropriate for research use.
  • White to off-white solid with defined storage recommendations.
  • Soluble in DMSO (100 mg/mL); ultrasonic assistance may be needed.
  • Available in small mg package sizes suitable for laboratory experiments.