Learn More
Medchemexpress LLC ω-hydroxy-DEET | 72236-22-7 | MFCD00871280 | 98.6% | 207.27 g/mol | C12H17NO2 | 5 MG
SDP
Catalog No. 5000222734
Encompass_Preferred
ω-Hydroxy-DEET (N,N-diethyl-3-(hydroxymethyl)benzamide) is a research-grade small molecule and a major metabolite of the insect repellent DEET, provided as a high-purity reference standard for biochemical and pharmacological studies. It is identified by CAS 72236-22-7, has formula C12H17NO2 and a molecular weight of 207.27 g/mol.
  • High purity (98.6%), suitable for analytical and assay work.
  • Soluble in DMSO at high concentration (≥100 mg/mL).
  • Liquid appearance, colorless to light yellow, with documented density.
  • Stable when stored protected from light; recommended cold storage for solutions.
  • Intended for research and analytical applications as a metabolite standard.
Catalog No. 50-002-22734 Supplier Medchemexpress LLC Supplier No. HY1366115MG
ω-Hydroxy-DEET (N,N-diethyl-3-(hydroxymethyl)benzamide) is a research-grade small molecule and a major metabolite of the insect repellent DEET, provided as a high-purity reference standard for biochemical and pharmacological studies. It is identified by CAS 72236-22-7, has formula C12H17NO2 and a molecular weight of 207.27 g/mol.
  • High purity (98.6%), suitable for analytical and assay work.
  • Soluble in DMSO at high concentration (≥100 mg/mL).
  • Liquid appearance, colorless to light yellow, with documented density.
  • Stable when stored protected from light; recommended cold storage for solutions.
  • Intended for research and analytical applications as a metabolite standard.