Learn More
Cayman Chemical Hydroxy DImetrIdazole 10mg
SDP
Catalog No. 502970746
Encompass_Preferred
A metabolite of dimetridazole, the nitroimidazole-based antibacterial and anticoccidial agent suspected to be carcinogenic and mutagenic to humans
Catalog No. 50-297-0746 Supplier Cayman Chemical Supplier No. 1620810 MG
A metabolite of dimetridazole, the nitroimidazole-based antibacterial and anticoccidial agent suspected to be carcinogenic and mutagenic to humans