Learn More
Cayman Chemical Hydroxy Dimetridazole, Purity: ≥98%, CAS: 936-05-0, 50 mg
SDP
Catalog No. 502970161
Encompass_Preferred
A metabolite of dimetridazole, the nitroimidazole-based antibacterial and anticoccidial agent suspected to be carcinogenic and mutagenic to humans
Catalog No. 50-297-0161 Supplier Cayman Chemical Supplier No. 1620850 MG
A metabolite of dimetridazole, the nitroimidazole-based antibacterial and anticoccidial agent suspected to be carcinogenic and mutagenic to humans