Learn More
Medchemexpress LLC HY-13863 10mg Medchemexpress, Hydroxy-Dynasore CAS:1256493-34-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1386310MG
Encompass_Preferred
Medchemexpress, HY-13863 10mg Hydroxy-Dynasore CAS:1256493-34-1 Hydroxy Dynasore (Dyngo-4a), a structural analog of Dynasore (HY-15304), is an potency improved, low cytotoxicity and non‐specific binding dynamin inhibitor with IC50 values of 0.38 μM and 2.3 μM for brain dynamin I and recombinant rat dynamin II, respectively. Hydroxy Dynasore inhibits dynamin‐dependent endocytosis of transferrin with an IC50 of 5.7 μM in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0485