Learn More
Hach Company Hydroxylamine Hydrochloride Solution, 100 g/L, 100 mL
Supplier:  Hach Company 81842
Encompass
For determination of volatile acids by the esterification method. Hach method 8196. 100 mL bottle
Catalog No. 50-199-4452