Learn More
Cayman Chemical Hydroxy Ipronidazole-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1156508-86-9, 2.5 mg
SDP
Catalog No. 502918353
Encompass_Preferred
An internal standard for the quantification of hydroxy ipronidazole by GC- or LC-MS
Catalog No. 50-291-8353 Supplier Cayman Chemical Supplier No. 391662.5 MG
An internal standard for the quantification of hydroxy ipronidazole by GC- or LC-MS