Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

βHydroxy-n-butyric Acid, Na Salt, MP Biomedicals

Catalog No. ICN210609 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN210609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN210609 Supplier MP Biomedicals, Inc Supplier No. 0521060905

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19054-57-0
Molecular Formula C4H7NaO3
Molecular Weight (g/mol) 126.087
InChI Key MOSCXNXKSOHVSQ-UHFFFAOYSA-M
Synonym sodium 2-hydroxybutanoate, sodium 2-hydroxybutyrate, sodium dl-2-hydroxybutyrate, 2-hydroxybutyric acid sodium salt, dl-2-hydroxybutyric acid sodium salt, sodium hydroxybutyrate, acmc-1btqj, sodium, a-hydroxybutyrate, sodium 2-oxidanylbutanoate, sodiumdl-2-hydroxybutyrate
PubChem CID 23663641
IUPAC Name sodium;2-hydroxybutanoate
SMILES CCC(C(=O)[O-])O.[Na+]

Specifications

Quantity 10 g
Formula Weight 126.09
Chemical Name or Material Beta-Hydroxy-N-Butyric Acid, Na Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.