Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxy naphthol blue disodium salt, For ACS analysis, MP Biomedicals™

Catalog No. ICN15252980 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15252980 100 g
ICN15252910 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15252980 Supplier MP Biomedicals, Inc Supplier No. 0215252980
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 165660-27-5
Molecular Formula C20H14N2Na2O11S3
Molecular Weight (g/mol) 600.495
InChI Key LXJHFXHZWYBBKJ-LUUCHEBKSA-N
Synonym `-(2-naphtholazo-3, 6-disulfonic acid)-2-naphthol-4-sulfonic acid trisodium salt, calcium indicator on sodium chloride
PubChem CID 131849366
IUPAC Name 3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonic acid;sodium
SMILES C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]

Specifications

Color Black to Blue
Quantity 100 g
Physical Form Powder
Formula Weight 598.5
Chemical Name or Material Hydroxy naphthol blue disodium salt, For ACS analysis
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.