Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Hydroxy Naphthol Blue, disodium salt, ACS reagent

Catalog No. AC204885000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC204885000 500 g Glass bottle
AC204880050 5 g Glass bottle
AC204881000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC204885000 Supplier Thermo Scientific Chemicals Supplier No. 204885000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 165660-27-5 | C20H12N2Na2O11S3 | 600.495 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165660-27-5
Molecular Formula C20H12N2Na2O11S3
Molecular Weight (g/mol) 600.495
MDL Number MFCD00004075
InChI Key LXJHFXHZWYBBKJ-LUUCHEBKSA-N
Synonym 2,2'-Dihydroxy-1,1'-azonaphthalene-3',4,6'-trisulfonic acid disodium salt
PubChem CID 131849366
IUPAC Name 3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonic acid;sodium
SMILES C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]

Specifications

Melting Point 274.0°C
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Formula Weight 598.47
Grade ACS Reagent
Packaging Glass bottle
Suitability (for calcium determination) passes
Color Blue to Purple
Quantity 500 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Hydroxy Naphthol Blue, disodium salt
Show More Show Less
Recommended Storage Room tempature

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.