Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxy Naphthol Blue Disodium Salt, Reagent, ACS,, Spectrum™ Chemical
SDP

Catalog No. 18603288 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-288 25 g Amber Glass Bottle
18-603-289 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-603-288 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H109225GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

H1092, 165660-27-5, C20H12O11N2S3Na2

Spectrum™ Chemical Hydroxy Naphthol Blue Disodium Salt, Reagent, ACS

Chemical Identifiers

CAS 165660-27-5
Molecular Formula C20H14N2Na2O11S3
Molecular Weight (g/mol) 600.49
InChI Key LXJHFXHZWYBBKJ-UHFFFAOYSA-N
IUPAC Name 3-hydroxy-4-[2-(2-oxo-4-sulfo-1,2-dihydronaphthalen-1-ylidene)hydrazin-1-yl]naphthalene-2,7-disulfonic acid disodium
SMILES [Na].[Na].OC1=C(NN=C2C(=O)C=C(C3=CC=CC=C23)S(O)(=O)=O)C2=CC=C(C=C2C=C1S(O)(=O)=O)S(O)(=O)=O

Specifications

CAS Max % 100%
Grade ACS
Packaging Amber Glass Bottle
Color Blue/Purple
Quantity 25 g
Chemical Name or Material Hydroxy Naphthol Blue Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.