Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxy Naphthol Blue Disodium Salt, Sigma-Aldrich

Catalog No. 01259037 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-037 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-259-037 Supplier MilliporeSigma Supelco Supplier No. 219916100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS reagent grade azo dye used as a pH indicator and for calcium determination.

Hydroxy naphthol blue disodium salt is a chemical compound belonging to the class of azo dyes commonly used as a pH indicator. It exists as small blue crystals. Hydroxy naphthol blue is suitable for the quantification of metal leaching in metal affinity chromatography. It changes color at pH range 12 to 13 from blue to reddish pink, in the presence of calcium ions and to deep blue in the presence of disodium ethylenediaminetetraacetic acid (EDTA).

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165660-27-5
Molecular Weight (g/mol) 598.49 g/mol

Specifications

Color Blue to Red
pH 7.9
Quantity 100 g
Grade ACS
Chemical Name or Material Hydroxy naphthol blue disodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.