Learn More
Medchemexpress LLC Hydroxy-PEG1-acid 10g | 89211-34-7 | 134.13 g/mol | C5H10O4 | 10 G
SDP
Catalog No. 5000227494
Encompass_Preferred
Hydroxy-PEG1-acid is a non-cleavable, single-unit polyethylene glycol (PEG) linker used as a short spacer in the synthesis of antibody-drug conjugates (ADCs). It has a terminal hydroxyl and carboxylic acid to enable common conjugation chemistries, is supplied as a purified research reagent (purity 95%), and is available in multiple pack sizes for laboratory use.
  • Provides a non-cleavable, single-unit PEG spacer for ADC linkages.
  • Terminal carboxylic acid enables standard amide coupling chemistries.
  • Hydroxyl group permits additional functionalization options.
  • High purity suitable for research applications (95%).
  • Available in multiple pack sizes, including a 10 g package.
Catalog No. 50-002-27494 Supplier Medchemexpress LLC Supplier No. HY11665510G
Add to Cart
Add to Cart
Hydroxy-PEG1-acid is a non-cleavable, single-unit polyethylene glycol (PEG) linker used as a short spacer in the synthesis of antibody-drug conjugates (ADCs). It has a terminal hydroxyl and carboxylic acid to enable common conjugation chemistries, is supplied as a purified research reagent (purity 95%), and is available in multiple pack sizes for laboratory use.
  • Provides a non-cleavable, single-unit PEG spacer for ADC linkages.
  • Terminal carboxylic acid enables standard amide coupling chemistries.
  • Hydroxyl group permits additional functionalization options.
  • High purity suitable for research applications (95%).
  • Available in multiple pack sizes, including a 10 g package.