Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydroxy-PEG2-acid | 1334286-77-9 | 97.0% | 178.18 g/mol | C7H14O5 | 5G
SDP

Catalog No. 5000235454
Encompass_Preferred

Hydroxy-PEG2-acid is a hydroxyl-terminated polyethylene glycol (PEG) linker intended for use in the synthesis of proteolysis-targeting chimeras (PROTACs). It provides a short, flexible spacer to connect ligands for target binding and E3 ligase recruitment, supporting conjugation chemistry in chemical biology and early drug discovery workflows.

  • Hydroxyl-terminated PEG2 linker
  • Designed for PROTAC synthesis and conjugation chemistry
  • CAS number 1334286-77-9
  • Molecular weight 178.18 g/mol
  • Chemical formula C7H14O5
  • Purity 97.0%
  • Available in multiple sizes from 100 mg to gram quantities

Catalog No. 50-002-35454 Supplier Medchemexpress LLC Supplier No. HY1243805G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Hydroxy-PEG2-acid is a hydroxyl-terminated polyethylene glycol (PEG) linker intended for use in the synthesis of proteolysis-targeting chimeras (PROTACs). It provides a short, flexible spacer to connect ligands for target binding and E3 ligase recruitment, supporting conjugation chemistry in chemical biology and early drug discovery workflows.

  • Hydroxyl-terminated PEG2 linker
  • Designed for PROTAC synthesis and conjugation chemistry
  • CAS number 1334286-77-9
  • Molecular weight 178.18 g/mol
  • Chemical formula C7H14O5
  • Purity 97.0%
  • Available in multiple sizes from 100 mg to gram quantities

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.