Learn More
Medchemexpress LLC Hydroxy-PEG4-acid 10g | 937188-59-5 | 10 G
SDP
Catalog No. 5000229240
Encompass_Preferred
Hydroxy-PEG4-acid is a four-unit polyethylene glycol (PEG) linker with a terminal hydroxyl and a carboxylic acid, supplied as a purified reagent for bioconjugation and linker synthesis. It is suitable for use as a non-cleavable spacer in antibody-drug conjugates (ADCs), and as a PEG-based linker in PROTACs and related chemistries.
  • Four-unit PEG spacer with terminal hydroxyl and carboxylic acid.
  • High purity suitable for research use.
  • Available in multiple pack sizes, including 10 G.
  • Stable when stored at recommended temperatures for long-term use.
  • Useful for ADC linker construction, PROTAC assembly, and general bioconjugation.
Catalog No. 50-002-29240 Supplier Medchemexpress LLC Supplier No. HY11710410G
Add to Cart
Add to Cart
Hydroxy-PEG4-acid is a four-unit polyethylene glycol (PEG) linker with a terminal hydroxyl and a carboxylic acid, supplied as a purified reagent for bioconjugation and linker synthesis. It is suitable for use as a non-cleavable spacer in antibody-drug conjugates (ADCs), and as a PEG-based linker in PROTACs and related chemistries.
  • Four-unit PEG spacer with terminal hydroxyl and carboxylic acid.
  • High purity suitable for research use.
  • Available in multiple pack sizes, including 10 G.
  • Stable when stored at recommended temperatures for long-term use.
  • Useful for ADC linker construction, PROTAC assembly, and general bioconjugation.