Learn More
Medchemexpress LLC Hydroxy-PEG4-acid 25g | 937188-59-5 | 266.29 g/mol | C11H22O7 | 25 G
SDP
Catalog No. 5000229239
Encompass_Preferred
Hydroxy-PEG4-acid is a PEG-based bifunctional linker with a terminal hydroxyl and a terminal carboxylic acid. It is designed for use as a non-cleavable four-unit PEG linker in conjugation chemistry, including antibody-drug conjugates (ADCs) and proteolysis-targeting chimeras (PROTACs).
  • Non-cleavable four-unit PEG linker suitable for ADC and PROTAC synthesis.
  • Terminal hydroxyl and carboxylic acid enable orthogonal functionalization.
  • Molecular weight 266.29 g/mol and molecular formula C11H22O7.
  • High purity (99.81%) for reliable conjugation performance.
  • Available in 25 G pack size for preparative applications.
Catalog No. 50-002-29239 Supplier Medchemexpress LLC Supplier No. HY11710425G
Hydroxy-PEG4-acid is a PEG-based bifunctional linker with a terminal hydroxyl and a terminal carboxylic acid. It is designed for use as a non-cleavable four-unit PEG linker in conjugation chemistry, including antibody-drug conjugates (ADCs) and proteolysis-targeting chimeras (PROTACs).
  • Non-cleavable four-unit PEG linker suitable for ADC and PROTAC synthesis.
  • Terminal hydroxyl and carboxylic acid enable orthogonal functionalization.
  • Molecular weight 266.29 g/mol and molecular formula C11H22O7.
  • High purity (99.81%) for reliable conjugation performance.
  • Available in 25 G pack size for preparative applications.