Learn More
Medchemexpress LLC Hydroxy-PEG4-CH2COOH | 70678-95-4 | 97.0% | C10H20O7 | 100 MG
SDP
Catalog No. 5000356662
Encompass_Preferred
Hydroxy-PEG4-CH2COOH is a PEG-based PROTAC linker essential for the synthesis of PROTACs. These molecules are designed with two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other binding to a specific target protein. This design enables the selective degradation of target proteins by harnessing the intracellular ubiquitin-proteasome system.
  • PEG-based linker
  • Essential for PROTAC synthesis
  • Features two distinct ligands connected by a linker
  • Targets an E3 ubiquitin ligase
  • Binds to a specific target protein
  • Enables selective degradation of target proteins
  • Utilizes the ubiquitin-proteasome system
Catalog No. 50-003-56662 Supplier Medchemexpress LLC Supplier No. HY140498100MG
Hydroxy-PEG4-CH2COOH is a PEG-based PROTAC linker essential for the synthesis of PROTACs. These molecules are designed with two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other binding to a specific target protein. This design enables the selective degradation of target proteins by harnessing the intracellular ubiquitin-proteasome system.
  • PEG-based linker
  • Essential for PROTAC synthesis
  • Features two distinct ligands connected by a linker
  • Targets an E3 ubiquitin ligase
  • Binds to a specific target protein
  • Enables selective degradation of target proteins
  • Utilizes the ubiquitin-proteasome system