Learn More
Medchemexpress LLC Hydroxy-PEG4-CH2COOH | 70678-95-4 | ≥97.0% | C10H20O7 | 500 MG
SDP
Catalog No. 5000356993
Encompass_Preferred
Hydroxy-PEG4-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase, and the other is for the target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-56993 Supplier Medchemexpress LLC Supplier No. HY140498500MG
Hydroxy-PEG4-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase, and the other is for the target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins