Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hydroxy-Peg4-T-Butyl Ester

Catalog No. 9025861G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-258-61G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-258-61G Supplier Sigma Aldrich Supplier No. 9025861G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This heterobifunctional, PEGylated crosslinker features a hydroxyl group at one end and t-butyl-protected carboxylic acid at the other, which can be deprotected with acidic conditions. The hydrophillic PEG linker facilitates solubility in biological applications. Hydroxy-PEG4-t-butyl ester can be used for bioconjugation or as a building block for synthesis of small molecules, conjugates of small molecules and/or biomolecules, or other tool compounds for chemical biology and medicinal chemistry that require ligation. Examples of applications include its synthetic incorporation into antibody-drug conjugates or proteolysis-targeting chimeras (PROTAC™ molecules) for targeted protein degradation.Recruiting cytotoxic T cells to folate-receptor-positive cancer cellsSyntheses and characterizations of novel pyrrolocoumarin probes for SNAP-tag labeling technologyHigh density orthogonal surface immobilization via photoactivated copper-free click chemistryPROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Specifications

CAS 14216-22-9
Molecular Formula C11H21ClN4O4
Linear Formula C11H21ClN4O4
Quantity 100 mg
Synonym tert-Butyl-1-hydroxy-3,6,9,12-tetraoxapentadecan-15-oate; HO-PEG4-CO-OtBu
Molecular Weight (g/mol) 308.76 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.