Learn More
Medchemexpress LLC HY-N6825 5mg Medchemexpress, Hydroxy-​α-​sanshool CAS:83883-10-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN68255MG
Encompass_Preferred
Medchemexpress, HY-N6825 5mg Hydroxy-​α-​sanshool CAS:83883-10-7 Hydroxy-​α-​sanshool is an alkylamide isolated from pepper, acts as a TRPA1 covalent and TRPV1 non-covalent agonist, with EC50s of 69 and 1.1 µM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0475