Learn More
Medchemexpress LLC [Hydroxy(tosyloxy)iodo]benzene | 27126-76-7 | 392.21 | 25 G
SDP
Catalog No. 5000403157
Encompass_Preferred
[Hydroxy(tosyloxy)iodo]benzene is a versatile reagent for the mild oxidation of aryl iodides at the iodine atom by ligand transfer. It is an effective reagent for the one-step conversion of ketones to the corresponding α-tosyloxy ketones. Additionally, it acts as a Brønsted acid and is expected to catalyze the enolization of ketones.
  • Versatile reagent for mild oxidation of aryl iodides.
  • Effective for one-step conversion of ketones to α-tosyloxy ketones.
  • Acts as a Brønsted acid.
  • Expected to catalyze the enolization of ketones.
Catalog No. 50-004-03157 Supplier Medchemexpress LLC Supplier No. 25GHY34619
[Hydroxy(tosyloxy)iodo]benzene is a versatile reagent for the mild oxidation of aryl iodides at the iodine atom by ligand transfer. It is an effective reagent for the one-step conversion of ketones to the corresponding α-tosyloxy ketones. Additionally, it acts as a Brønsted acid and is expected to catalyze the enolization of ketones.
  • Versatile reagent for mild oxidation of aryl iodides.
  • Effective for one-step conversion of ketones to α-tosyloxy ketones.
  • Acts as a Brønsted acid.
  • Expected to catalyze the enolization of ketones.