Learn More
Medchemexpress LLC Hydroxyacetone | 116-09-6 | MFCD00004669 | 25g
SDP
Catalog No. 5000267217
Encompass_Preferred
Hydroxyacetone is a dehydration product of glycerol Hydroxyacetone is a platform for the electrocatalytic synthesis of acetone 1 2-propanediol and 2-propanol Hydroxyacetone can also be used as a glycerol substitute for electrocatalytic hydrogenation and hydrodeoxygenation processes[1]
Catalog No. 50-002-67217 Supplier Medchemexpress LLC Supplier No. HYY136625G
Hydroxyacetone is a dehydration product of glycerol Hydroxyacetone is a platform for the electrocatalytic synthesis of acetone 1 2-propanediol and 2-propanol Hydroxyacetone can also be used as a glycerol substitute for electrocatalytic hydrogenation and hydrodeoxygenation processes[1]