Learn More
Medchemexpress LLC Hydroxyacetone | 116-09-6 | MFCD00004669 | ≥98.0% | 5g
SDP
Catalog No. 5000267106
Encompass_Preferred
Hydroxyacetone is a dehydration product of glycerol Hydroxyacetone is a platform for the electrocatalytic synthesis of acetone 1 2-propanediol and 2-propanol Hydroxyacetone can also be used as a glycerol substitute for electrocatalytic hydrogenation and hydrodeoxygenation processes[1]
Catalog No. 50-002-67106 Supplier Medchemexpress LLC Supplier No. HYY13665G
Hydroxyacetone is a dehydration product of glycerol Hydroxyacetone is a platform for the electrocatalytic synthesis of acetone 1 2-propanediol and 2-propanol Hydroxyacetone can also be used as a glycerol substitute for electrocatalytic hydrogenation and hydrodeoxygenation processes[1]